Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3,5-Trimethoxybenzene

Catalog No. 138827250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-882-7250G 250 g
13-882-750G 50 g
13-882-710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-882-7250G Supplier Sigma Aldrich Supplier No. 138827250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3,5-Trimethoxybenzene

Specifications

CAS 621-23-8
Melting Point 50°C to 53°C (lit.)
Boiling Point 255°C (lit.)
Molecular Formula C9H12O3
Linear Formula C6H3(OCH3)3
MDL Number MFCD00008385
Quantity 250 g
Synonym Phloroglucinol trimethyl ether
Molecular Weight (g/mol) 168.19
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.