Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorosulfonyl isocyanate, 98+%

Catalog No. AC162040010
Change view
Click to view available options
Quantity:
5 mL
50 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162040010 1 L Glass bottle
AC162040050 5 mL Glass bottle
AC162040500 50 mL Glass bottle
AC162042500 250 mL Glass bottle
AC162040025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC162040010 Supplier Thermo Scientific Chemicals Supplier No. 162040010
Only null left
Add to Cart

CAS: 1189-71-5 | CClNO3S | 141.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent, C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189-71-5
Molecular Formula CClNO3S
Molecular Weight (g/mol) 141.53
InChI Key WRJWRGBVPUUDLA-UHFFFAOYSA-N
Synonym chlorosulfonyl isocyanate, chlorosulfonylisocyanate, sulfuryl chloride isocyanate, n-carbonylsulfamyl chloride, chlorosulphonyl isocyanate, sulfurisocyanatidic chloride, n-chlorosulfonyl isocyanate, chlorosulfonyl imino methanone, csi, sulfurisocyanatidoyl chloride
PubChem CID 70918
IUPAC Name N-(oxomethylidene)sulfamoyl chloride
SMILES C(=NS(=O)(=O)Cl)=O

Specifications

Melting Point -44.0°C
Color White
Density 1.6260g/mL
Boiling Point 106.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Refractive Index 1.4450 to 1.4490
Linear Formula ClSO2NCO
Quantity 1 L
Fieser 01,117; 02,70; 03,51; 04,90; 05,132; 06,122; 07,65; 08,105; 10,94; 11,125; 12,122; 13,80
Specific Gravity 1.626
Solubility Information Solubility in water: reacts. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons,miscible with saturated and chlorinated,hydrocarbons
Viscosity 0.996 mPa.s (20°C)
Formula Weight 141.53
Percent Purity 98+%
Physical Form Crystalline Powder, Crystals or Granules
Chemical Name or Material Chlorosulfonyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 214-715-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.