Learn More
Medchemexpress LLC 1,3,5-trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene | 1709-70-2 | 99.0% | 100 G
SDP
Catalog No. 5000234130
Encompass_Preferred
Irganox 1330 is a sterically hindered phenolic antioxidant used to protect polymers from thermal and oxidative degradation during processing and service. It is used as an additive in polyolefins to improve thermal stability and extend service life.
  • Provides thermal and oxidative stabilization for polymers.
  • Effective in polyethylene, polypropylene, LLDPE, and HDPE formulations.
  • Powder form enables easy dispersion during compounding.
  • High purity for consistent antioxidant performance.
  • Molecular weight 775.20 g/mol and formula C54H78O3.
Catalog No. 50-002-34130 Supplier Medchemexpress LLC Supplier No. HY120388100G
Irganox 1330 is a sterically hindered phenolic antioxidant used to protect polymers from thermal and oxidative degradation during processing and service. It is used as an additive in polyolefins to improve thermal stability and extend service life.
  • Provides thermal and oxidative stabilization for polymers.
  • Effective in polyethylene, polypropylene, LLDPE, and HDPE formulations.
  • Powder form enables easy dispersion during compounding.
  • High purity for consistent antioxidant performance.
  • Molecular weight 775.20 g/mol and formula C54H78O3.