Learn More
Medchemexpress LLC 1,3,5-trimethyl-2-(4-methylphenyl)sulfonylbenzene | 5184-64-5 | 99.8% | 274.38 g/mol | C16H18O2S | 1 ML
SDP
Catalog No. 5000228120
Encompass_Preferred
ESI-05 (NSC 116966) is a selective small-molecule inhibitor of exchange protein directly activated by cAMP 2 (EPAC2). It inhibits cAMP-mediated EPAC2 guanine nucleotide exchange factor (GEF) activity (reported IC50 ≈ 0.43 μM). The product is provided as a 10 mM solution in DMSO (1 mL) and is available as a solid; reported purity is ~99.78%.
  • Selective EPAC2 inhibitor with reported IC50 ≈ 0.43 μM.
  • Supplied as 10 mM solution in DMSO, 1 mL ready for use.
  • High purity (~99.8%) suitable for research applications.
  • Also available as solid in multiple sizes for flexibility.
  • Applicable to studies of insulin secretion, diabetes, and neuronal signaling.
Catalog No. 50-002-28120 Supplier Medchemexpress LLC Supplier No. HY11765610MM 1ML
ESI-05 (NSC 116966) is a selective small-molecule inhibitor of exchange protein directly activated by cAMP 2 (EPAC2). It inhibits cAMP-mediated EPAC2 guanine nucleotide exchange factor (GEF) activity (reported IC50 ≈ 0.43 μM). The product is provided as a 10 mM solution in DMSO (1 mL) and is available as a solid; reported purity is ~99.78%.
  • Selective EPAC2 inhibitor with reported IC50 ≈ 0.43 μM.
  • Supplied as 10 mM solution in DMSO, 1 mL ready for use.
  • High purity (~99.8%) suitable for research applications.
  • Also available as solid in multiple sizes for flexibility.
  • Applicable to studies of insulin secretion, diabetes, and neuronal signaling.