Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3,5-Trimethylpyrazole | 1072-91-9 | MFCD00015536 | 500 MG
SDP

Catalog No. 5000420184
Encompass_Preferred

1,3,5-Trimethylpyrazole is a compound primarily used for chemical synthesis.

  • High purity of 99.91%
  • Molecular weight of 110.16
  • Chemical formula C6H10N2
  • Appearance as a solid below 38°C and liquid above 41°C
  • Colorless to off-white appearance
  • Soluble in DMSO and H2O at 100 mg/mL, requiring ultrasonic for dissolution
  • Multiple in vivo dissolution protocols available, including those with DMSO, PEG300, Tween-80, Saline, SBE-β-CD, and Corn Oil
  • Long-term storage recommendations: -20°C for 3 years, 4°C for 2 years for pure form; -80°C for 6 months, -20°C for 1 month for in solvent

Catalog No. 50-004-20184 Supplier Medchemexpress LLC Supplier No. HYN7086500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3,5-Trimethylpyrazole is a compound primarily used for chemical synthesis.

  • High purity of 99.91%
  • Molecular weight of 110.16
  • Chemical formula C6H10N2
  • Appearance as a solid below 38°C and liquid above 41°C
  • Colorless to off-white appearance
  • Soluble in DMSO and H2O at 100 mg/mL, requiring ultrasonic for dissolution
  • Multiple in vivo dissolution protocols available, including those with DMSO, PEG300, Tween-80, Saline, SBE-β-CD, and Corn Oil
  • Long-term storage recommendations: -20°C for 3 years, 4°C for 2 years for pure form; -80°C for 6 months, -20°C for 1 month for in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.