Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Tris(bromomethyl)benzene 98.0+%, TCI America™
SDP

Catalog No. T27191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27191G 1 g
T27195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27191G Supplier TCI America Supplier No. T27191G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18226-42-1
Molecular Formula C9H9Br3
Molecular Weight (g/mol) 356.883
MDL Number MFCD00182500
InChI Key GHITVUOBZBZMND-UHFFFAOYSA-N
PubChem CID 530258
IUPAC Name 1,3,5-tris(bromomethyl)benzene
SMILES C1=C(C=C(C=C1CBr)CBr)CBr

Specifications

Melting Point 99°C
Quantity 1 g
UN Number 3261
Synonym alpha,alpha′C,alpha′C′C-Tribromomesitylene
Formula Weight 356.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tris(bromomethyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.