Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Trithiane

Catalog No. AC140521000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC140521000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC140521000 Supplier Thermo Scientific Chemicals Supplier No. 140521000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

CAS: 291-21-4 | C3H6S3 | 138.26 g/mol

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 291-21-4
Molecular Formula C3H6S3
Molecular Weight (g/mol) 138.26
InChI Key LORRLQMLLQLPSJ-UHFFFAOYSA-N
IUPAC Name 1,3,5-trithiane
SMILES C1SCSCS1

Specifications

Melting Point 214.0°C to 215.0°C
Quantity 100 g
Beilstein 19, 382
Fieser 03,329; 04,564
Solubility Information Solubility aq. soln.: insoluble.
Formula Weight 138.27
Chemical Name or Material 1,3,5-Trithiane
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

EINECSNumber 206-029-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.