Learn More
Cayman Chemical 1-(3,5-Dimethylphenyl)piperazine, Purity: ≥98%, CAS: 105907-65-1, 500 mg
SDP
Catalog No. 502984738
Encompass_Preferred
A piperidine compound that is inactive at dopamine and serotonin receptors that may be used as a heterocyclic building block
Catalog No. 50-298-4738 Supplier Cayman Chemical Supplier No. 20627500 MG
A piperidine compound that is inactive at dopamine and serotonin receptors that may be used as a heterocyclic building block