Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3,(6,7)-Naphthalenetrisulfonic acid trisodium salt hydrate

Catalog No. 3107431KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-074-31KG 1 kg
31-074-3250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-074-31KG Supplier Sigma Aldrich Supplier No. 3107431KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3,(6,7)-Naphthalenetrisulfonic acid trisodium salt hydrate is an anionic chromophore.

Specifications

CAS 123409-01-8
Molecular Formula C10H5Na3O9S3 -+ xH2O
Linear Formula C10H5(SO3Na)3 -+ xH2O
MDL Number MFCD00149263
Quantity 1 kg
Molecular Weight (g/mol) 434.31 (anhydrous basis)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.