Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,6,8-Tetrabromopyrene 98.0+%, TCI America™
SDP

Catalog No. T27165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T27165G Supplier TCI America Supplier No. T27165G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 128-63-2
Molecular Formula C16H6Br4
Molecular Weight (g/mol) 517.84
MDL Number MFCD00428682
InChI Key ZKBKRTZIYOKNRG-UHFFFAOYSA-N
PubChem CID 67188
IUPAC Name 1,3,6,8-tetrabromopyrene
SMILES BrC1=CC(Br)=C2C=CC3=C(Br)C=C(Br)C4=CC=C1C2=C34

Specifications

Quantity 5 g
Formula Weight 517.84
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,6,8-Tetrabromopyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.