Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,6,8-Tetranitrocarbazole (wetted with ca. 40% Water) (unit weight on dry weight basis) 70.0+%, TCI America™
SDP

Catalog No. T015925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T015925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T015925G Supplier TCI America Supplier No. T015925G

Chemical Identifiers

CAS 4543-33-3
Molecular Formula C12H5N5O8
Molecular Weight (g/mol) 347.199
MDL Number MFCD00022211
InChI Key JUSWGNJYSBSOFM-UHFFFAOYSA-N
PubChem CID 72755
IUPAC Name 1,3,6,8-tetranitro-9H-carbazole
SMILES C1=C(C=C2C3=CC(=CC(=C3NC2=C1[N+](=O)[O-])[N+](=O)[O-])[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 291°C
Quantity 25 g
UN Number 1325
Formula Weight 347.20
Percent Purity ≥70.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,6,8-Tetranitrocarbazole (wetted with ca. 40% Water) (unit weight on dry weight basis)
RTECSNumber FE6325000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.