Learn More
Cayman Chemical 1,3,6,8-Pyrenetetrasulfonic Acid (sodium salt hydrate), Purity: ≥95%, CAS: 1771776-02-3, 1 g
SDP
Catalog No. 502989724
Encompass_Preferred
A synthetic intermediate; has been used to stabilize intermolecular interactions for the crystallization of L. mexicana pyruvate kinase
Catalog No. 50-298-9724 Supplier Cayman Chemical Supplier No. 299801 G
A synthetic intermediate; has been used to stabilize intermolecular interactions for the crystallization of L. mexicana pyruvate kinase