Learn More
TARGETMOL CHEMICALS INC 1 3 6-Tri-O-galloyl-beta-D-glu
SDP
Catalog No. 502877168
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 1,3,6-Tri-O-galloyl-beta-D-glucose (1,3,6-Tri-O-galloylglucose) is a phenolic compound found in Black Walnut Kernels, exhibiting anti-inflammatory activity. Purity 99.37%
Catalog No. 50-287-7168 Supplier TARGETMOL CHEMICALS INC Supplier No. TN11661MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 1,3,6-Tri-O-galloyl-beta-D-glucose (1,3,6-Tri-O-galloylglucose) is a phenolic compound found in Black Walnut Kernels, exhibiting anti-inflammatory activity. Purity 99.37%