Learn More
Medchemexpress LLC 1,3,6-trihydroxy-2-(3-hydroxy-3-methylbutyl)-7-methoxy-8-(3-methylbut-2-enyl)xanthen-9-one | 149155-01-1 | MFCD29904529 | 95.0% | 428.47 | C24H28O7 | 10 MG
SDP
Catalog No. 5000289744
Encompass_Preferred
Cratoxylone is a natural xanthone derivative isolated from the bark of Cratoxylum cochinchinense. It exhibits antiplasmodial activity and is supplied as research-grade material for biochemical and antiparasitic studies. Typical analytical data reported by the manufacturer include CAS 149155-01-1, molecular formula C24H28O7, molecular weight 428.47, and purity around 95.0%. Storage recommendations are sealed, away from moisture and light, and in solvent: -80°C for up to 6 months or -20°C for up to 1 month.
  • Natural xanthone derivative with antiplasmodial activity.
  • Research-grade chemical with reported purity of 95.0%.
  • Chemical formula C24H28O7 and molecular weight 428.47.
  • Available in small milligram pack sizes, including 10 MG.
  • Suitable for biochemical and antiparasitic research applications.
  • Store sealed, protected from moisture and light; follow low-temperature solvent storage guidelines.
Catalog No. 50-002-89744 Supplier Medchemexpress LLC Supplier No. HYN625110MG
Cratoxylone is a natural xanthone derivative isolated from the bark of Cratoxylum cochinchinense. It exhibits antiplasmodial activity and is supplied as research-grade material for biochemical and antiparasitic studies. Typical analytical data reported by the manufacturer include CAS 149155-01-1, molecular formula C24H28O7, molecular weight 428.47, and purity around 95.0%. Storage recommendations are sealed, away from moisture and light, and in solvent: -80°C for up to 6 months or -20°C for up to 1 month.
  • Natural xanthone derivative with antiplasmodial activity.
  • Research-grade chemical with reported purity of 95.0%.
  • Chemical formula C24H28O7 and molecular weight 428.47.
  • Available in small milligram pack sizes, including 10 MG.
  • Suitable for biochemical and antiparasitic research applications.
  • Store sealed, protected from moisture and light; follow low-temperature solvent storage guidelines.