Learn More
Cayman Chemical 1,3,7-Trimethyluric Acid, Purity: ≥98%, CAS: 5415-44-1, 50 mg
SDP
Catalog No. 502966982
Encompass_Preferred
A metabolite of caffeine; scavenges hydroxyl radicals in a cell-free assay at 500 µM; inhibits t-butyl hydroperoxide-induced lipid peroxidation by 56.5% in isolated human erythrocyte membranes
Catalog No. 50-296-6982 Supplier Cayman Chemical Supplier No. 1694950 MG
A metabolite of caffeine; scavenges hydroxyl radicals in a cell-free assay at 500 µM; inhibits t-butyl hydroperoxide-induced lipid peroxidation by 56.5% in isolated human erythrocyte membranes