Learn More
Medchemexpress LLC 1,3,8-Triazaspiro[4.5]decan-4-one, 8-[(5-chloro-2-thienyl)sulfonyl]-1-phenyl- | 1023043-30-2 | 99.6% | 411.93 | 25 MG
SDP
Catalog No. 5000439112
Encompass_Preferred
ATP synthase inhibitor 1 is a potent inhibitor of the c subunit of the F1/FO-ATP synthase complex. It inhibits mitochondrial permeability transition pore (mPTP) opening but does not affect ATP levels.
  • Potent inhibitor of c subunit of the F1/FO-ATP synthase complex
  • Inhibits mitochondrial permeability transition pore (mPTP) opening
  • Does not affect ATP levels
Catalog No. 50-004-39112 Supplier Medchemexpress LLC Supplier No. 25MGHY112715
ATP synthase inhibitor 1 is a potent inhibitor of the c subunit of the F1/FO-ATP synthase complex. It inhibits mitochondrial permeability transition pore (mPTP) opening but does not affect ATP levels.
  • Potent inhibitor of c subunit of the F1/FO-ATP synthase complex
  • Inhibits mitochondrial permeability transition pore (mPTP) opening
  • Does not affect ATP levels