Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3,8-triazaspiro[4.5]decane-2,4-dione

Catalog No. CDS012186 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS012186 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS012186 Supplier Sigma Aldrich Supplier No. CDS012186100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,3-dibromotetrafluorobenzene is a colorless liquid
  • 1,3-dibromotetrafluorobenzene crystallizes with 4,4α-dipyridyl to form the noncovalent copolymers.1,3-dibromotetrafluorobenzene was used to prepare starting compounds for the synthesis of fluorinated liquid crystal derivatives
  • It was also used in the synthesis of halogen bonded adducts by undergoing in situ cryocrystallization with various oxygenated solvents

Specifications

Molecular Formula C7H11N3O2
Linear Formula C7H11N3O2
Quantity 1 g
MDL Number MFCD06660736
Molecular Weight (g/mol) 169.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.