Learn More
Jackson Hirsh Inc 1 3/8X4 1/4 LAMINTNG POU 500PK
Catalog No. NC0307475
Encompass
  • Pouch dimensions: 1 3/8 in x 4 1/4 in
  • Pack quantity: 500 pouches
  • Total thickness: 14 mil
  • Thickness per side: 7 mil
  • Finish: Clear
  • Feature: Pre-punched slot on short side
  • Material: High-quality plastic
  • Intended use: Label and tag lamination
Catalog No. NC0307475 Supplier Jackson Hirsh Inc Supplier No. LABEL 20WM2
  • Pouch dimensions: 1 3/8 in x 4 1/4 in
  • Pack quantity: 500 pouches
  • Total thickness: 14 mil
  • Thickness per side: 7 mil
  • Finish: Clear
  • Feature: Pre-punched slot on short side
  • Material: High-quality plastic
  • Intended use: Label and tag lamination