Learn More
AdipoGen 1 3-Acetonedicarboxylic acid
SDP
Catalog No. 502559895
Encompass_Preferred
Chemical. CAS 542-05-2. Formula C5H6O5. Molecular Weight 146.1. Acetonedicarboxylic acid is primarily used as building blocks in organic chemistry, particularly in the synthesis of heterocyclic rings and in the Weiss-Cook reaction.
Catalog No. 50-255-9895 Supplier AdipoGen Supplier No. CDXY0346G100
Chemical. CAS 542-05-2. Formula C5H6O5. Molecular Weight 146.1. Acetonedicarboxylic acid is primarily used as building blocks in organic chemistry, particularly in the synthesis of heterocyclic rings and in the Weiss-Cook reaction.