Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-acetonedicarboxylic acid, 96%

Catalog No. p-3756521
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173151000 100 g Glass Bottle
AC173155000 500 g Glass Bottle
AC173150025 2.5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC173151000 Supplier Thermo Scientific Chemicals Supplier No. 173151000
Only null left
Add to Cart

CAS: 542-05-2 | C5H6O5 | 146.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-05-2
Molecular Formula C5H6O5
Molecular Weight (g/mol) 146.1
MDL Number MFCD00002711
InChI Key OXTNCQMOKLOUAM-UHFFFAOYSA-N
Synonym 1,3-acetonedicarboxylic acid, 3-oxoglutaric acid, acetonedicarboxylic acid, pentanedioic acid, 3-oxo, acetone-1,3-dicarboxylic acid, 3-ketoglutaric acid, unii-ih7p7wo21p, 1,3-acetonedicarboxylic acid; 3-ketoglutaric acid, ih7p7wo21p, acetondicarbonsaure
PubChem CID 68328
IUPAC Name 3-oxopentanedioic acid
SMILES C(C(=O)CC(=O)O)C(=O)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 133°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass Bottle
Linear Formula HO2CCH2COCH2CO2H
Quantity 100 g
Beilstein 03,789
Fieser 11,2
Merck Index 15,67
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in ethyl acetate and ether,insoluble in benzene,chloroform,ligroin,decomposes in hot water,acids,alkalies to co2 an,d acetone
Formula Weight 146.1
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 1, 3-Acetonedicarboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective cloth
EINECSNumber 208-797-9
Recommended Storage preferably stored under cool conditions

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.