Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Allyl-4-hydroxyphenyl)ethan-1-one, 97%, Thermo Scientific™

Catalog No. KM05715EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
KM05715EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM05715EA Supplier Thermo Scientific Chemicals Supplier No. KM05715EA

Chemical Identifiers

CAS 1132-05-4
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00068015
InChI Key XVTCWUFLNLZPEJ-UHFFFAOYSA-N
PubChem CID 259653
IUPAC Name 1-(4-hydroxy-3-prop-2-enylphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)O)CC=C

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 3'-allyl-4'-hydroxyacetophenone, acetophenone, 3'-allyl-4'-hydroxy, 1-3-allyl-4-hydroxyphenyl ethan-1-one, 1-3-allyl-4-hydroxyphenyl ethanone, ethanone, 1-4-hydroxy-3-2-propenyl phenyl, 1-4-hydroxy-3-prop-2-en-1-yl phenyl ethanone, ethanone,1-4-hydroxy-3-2-propen-1-yl phenyl, ethanone, 1-4-hydroxy-3-2-propen-1-yl phenyl, zlchem 1234
Formula Weight 176.21
Percent Purity 95%
Chemical Name or Material 1-(3-Allyl-4-hydroxyphenyl)ethan-1-one
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.