Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Aminopropyl)-2-methylpiperidine 98.0+%, TCI America™
SDP

Catalog No. A083325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A083325ML 25 mL
A0833500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A083325ML Supplier TCI America Supplier No. A083325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25560-00-3
Molecular Formula C9H20N2
Molecular Weight (g/mol) 156.273
MDL Number MFCD00006517
InChI Key YYAYTNPNFKPFNG-UHFFFAOYSA-N
Synonym 1-(3-Aminopropyl)-2-pipecoline
PubChem CID 520211
IUPAC Name 3-(2-methylpiperidin-1-yl)propan-1-amine
SMILES CC1CCCCN1CCCN

Specifications

Color Yellow
Boiling Point 170°C
Quantity 25 mL
UN Number 3267
Formula Weight 156.27
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(3-Aminopropyl)-2-methylpiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.