Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Aminopropyl)-4-methylpiperazine, 98%

Catalog No. AAL0487603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0487603 1 g
AAL0487606 5 g
AAL0487614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0487603 Supplier Thermo Scientific Chemicals Supplier No. L0487603
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4572-03-6 | C8H19N3 | 157.26 g/mol

1-(3-Aminopropyl)-4-methylpiperazine is used as a precursor in organic synthesis for the preparation of furan-2-carboxylic acid [3-(4-methyl-piperazin-1-yl)-propyl]-amide by reacting with furan-2-carbonyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Aminopropyl)-4-methylpiperazine is used as a precursor in organic synthesis for the preparation of furan-2-carboxylic acid [3-(4-methyl-piperazin-1-yl)-propyl]-amide by reacting with furan-2-carbonyl chloride.

Notes
Air sensitive. Keep in a dry, cool and well-ventilated place. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4572-03-6
Molecular Formula C8H19N3
Molecular Weight (g/mol) 157.26
MDL Number MFCD00014616
InChI Key RGUABPVONIGVAT-UHFFFAOYSA-N
Synonym 3-4-methylpiperazin-1-yl propan-1-amine, 1-3-aminopropyl-4-methylpiperazine, 1-piperazinepropanamine, 4-methyl, 3-4-methyl-1-piperazinyl propylamine, 4-3-aminopropyl-1-methylpiperazine, 1-methyl-4-3'-aminopropyl piperazine, 4-methyl-1-piperazinepropanamine, 3-4-methyl-piperazin-1-yl-propylamine, 3-4-methylpiperazin-1-yl propylamine, n-methyl-n'-3-aminopropyl piperazine
PubChem CID 78328
IUPAC Name 3-(4-methylpiperazin-1-yl)propan-1-amine
SMILES CN1CCN(CC1)CCCN

Specifications

Density 0.924
Boiling Point 113°C to 114°C (13 mmHg)
Refractive Index 1.482
Quantity 1 g
UN Number UN2735
Beilstein 105964
Sensitivity Air Sensitive
Formula Weight 157.26
Percent Purity 98%
Chemical Name or Material 1-(3-Aminopropyl)-4-methylpiperazine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 224-954-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.