Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(3-Aminopropyl)imidazole

Catalog No. 272264250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-226-4250G 250 g
27-226-450G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 27-226-4250G Supplier Sigma Aldrich Supplier No. 272264250G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-(3-aminopropyl)imidazole was used in the synthesis of pH-sensitive polyaspartamide derivatives
  • It was also used in the preparation of pH-sensitive amphiphilic polymers with different degrees of octaceylamine (c18) substitution by graft reaction

Specifications

CAS 5036-48-6
Density 1.049 g/mL (at 25°C (literature))
Molecular Formula C6H11N3
Refractive Index n20/D 1.519 (literature)
Linear Formula C6H11N3
MDL Number MFCD00009819
Quantity 250 g
Molecular Weight (g/mol) 125.17
Percent Purity ≥97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.