Learn More
Medchemexpress LLC 1,3-benzenedicarboxaldehyde, 2,4,6-trihydroxy- | 4396-13-8 | MFCD24674149 | 99.6% | 182.13 g·mol⁻¹ | C8H6O5 | 250 MG
SDP
Catalog No. 5000282942
Encompass_Preferred
Diformylphloroglucinol is a phloroglucinol derivative used as a synthetic intermediate in organic and natural product synthesis and for research chemistry applications. It is supplied as a pink to red solid with high purity suitable for synthetic and analytical workflows.
  • High purity (99.6% HPLC) suitable for synthesis and analysis.
  • Pink to red solid, easy to handle and weigh.
  • Molecular formula C8H6O5 and molecular weight 182.13 g·mol⁻¹.
  • Useful as a building block in organic and natural product synthesis.
  • Provided with a certificate of analysis for batch verification.
Catalog No. 50-002-82942 Supplier Medchemexpress LLC Supplier No. HYW130232250MG
Diformylphloroglucinol is a phloroglucinol derivative used as a synthetic intermediate in organic and natural product synthesis and for research chemistry applications. It is supplied as a pink to red solid with high purity suitable for synthetic and analytical workflows.
  • High purity (99.6% HPLC) suitable for synthesis and analysis.
  • Pink to red solid, easy to handle and weigh.
  • Molecular formula C8H6O5 and molecular weight 182.13 g·mol⁻¹.
  • Useful as a building block in organic and natural product synthesis.
  • Provided with a certificate of analysis for batch verification.