Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzenedimethanethiol 98.0+%, TCI America™
SDP

Catalog No. B38541G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38541G 1 g
B38545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38541G Supplier TCI America Supplier No. B38541G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 41563-69-3
Molecular Formula C8H10S2
Molecular Weight (g/mol) 170.288
MDL Number MFCD00012311
InChI Key JSNABGZJVWSNOB-UHFFFAOYSA-N
PubChem CID 586424
IUPAC Name [3-(sulfanylmethyl)phenyl]methanethiol
SMILES C1=CC(=CC(=C1)CS)CS

Specifications

Color Yellow
Boiling Point 170°C
Quantity 1 g
Synonym 1,3-Bis(mercaptomethyl)benzene, alpha,alpha′C-Dimercapto-m-xylene
Formula Weight 170.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Benzenedimethanethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.