Learn More
Medchemexpress LLC 1,3-benzenediol, 4-butyl- | 18979-61-8 | MFCD01684800 | 99.5% | 166.22 g·mol⁻¹ | C10H14O2 | 50 MG
SDP
Catalog No. 5000240714
Encompass_Preferred
4-Butylresorcinol is a phenolic research reagent used to study melanogenesis and enzymatic oxidation. It inhibits tyrosinase (IC50 = 11.27 μM) and reduces melanin synthesis in cellular and cell-free assays. Supplied as a white solid for laboratory use; for research only.
  • Tyrosinase inhibitor with documented potency (IC50 = 11.27 μM).
  • Reduces melanin synthesis in cell and cell-free systems.
  • High reported purity, suitable for analytical and biological studies.
  • Molecular weight 166.22 g·mol⁻¹; formula C10H14O2.
  • Supplied as a white solid for laboratory research use only.
Catalog No. 50-002-40714 Supplier Medchemexpress LLC Supplier No. HY10736950MG
4-Butylresorcinol is a phenolic research reagent used to study melanogenesis and enzymatic oxidation. It inhibits tyrosinase (IC50 = 11.27 μM) and reduces melanin synthesis in cellular and cell-free assays. Supplied as a white solid for laboratory use; for research only.
  • Tyrosinase inhibitor with documented potency (IC50 = 11.27 μM).
  • Reduces melanin synthesis in cell and cell-free systems.
  • High reported purity, suitable for analytical and biological studies.
  • Molecular weight 166.22 g·mol⁻¹; formula C10H14O2.
  • Supplied as a white solid for laboratory research use only.