Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-benzenediol, 5-[(1E)-2-phenylethenyl]- | 22139-77-1 | MFCD00210544 | 99.9% | 212.24 | C14H12O2 | 10 MG
SDP

Catalog No. 5000289649
Encompass_Preferred

Pinosylvin is a stilbenoid compound and resveratrol analogue reported to have antibacterial activity and to induce apoptosis and autophagy in cell-based studies. Supplied as a white to off-white solid for research use only; handle according to safety guidance and protect from light.

  • Cas number: 22139-77-1
  • Molecular formula: C14H12O2
  • Molecular weight: 212.24
  • Purity: 99.94%
  • Appearance: white to off-white solid
  • Storage: store at 4°C, protect from light; in solution, -80°C up to 6 months, -20°C up to 1 month
  • Available forms: solid (5 mg, 10 mg, 25 mg, 50 mg, 100 mg), and 10 mM solution in DMSO
  • Intended use: for research use only, not for human therapeutic applications

Catalog No. 50-002-89649 Supplier Medchemexpress LLC Supplier No. HYN238710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pinosylvin is a stilbenoid compound and resveratrol analogue reported to have antibacterial activity and to induce apoptosis and autophagy in cell-based studies. Supplied as a white to off-white solid for research use only; handle according to safety guidance and protect from light.

  • Cas number: 22139-77-1
  • Molecular formula: C14H12O2
  • Molecular weight: 212.24
  • Purity: 99.94%
  • Appearance: white to off-white solid
  • Storage: store at 4°C, protect from light; in solution, -80°C up to 6 months, -20°C up to 1 month
  • Available forms: solid (5 mg, 10 mg, 25 mg, 50 mg, 100 mg), and 10 mM solution in DMSO
  • Intended use: for research use only, not for human therapeutic applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.