Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-benzenediol, 5-methyl-, hydrate (1:1) | 6153-39-5 | MFCD00149092 | 95.0% | 142.15 | C7H10O3 | 1 ML
SDP

Catalog No. 5000300016
Encompass_Preferred

5-Methylresorcinol monohydrate is the monohydrate form of 5-methylresorcinol supplied for research use. It functions as a tyrosinase inhibitor and is provided as both solid material and prepared solutions for biochemical and cell-based studies.

  • Monohydrate form of 5-methylresorcinol.
  • Acts as a tyrosinase inhibitor suitable for biochemical assays.
  • Available as solid and as a 10 mM solution in DMSO.
  • Purity listed as 95.0%.
  • Molecular formula C7H10O3; molecular weight 142.15 g/mol.
  • Solubility ≥ 2.5 mg/mL (≈17.59 mM) in recommended solvents.

Catalog No. 50-003-00016 Supplier Medchemexpress LLC Supplier No. HYW01571510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methylresorcinol monohydrate is the monohydrate form of 5-methylresorcinol supplied for research use. It functions as a tyrosinase inhibitor and is provided as both solid material and prepared solutions for biochemical and cell-based studies.

  • Monohydrate form of 5-methylresorcinol.
  • Acts as a tyrosinase inhibitor suitable for biochemical assays.
  • Available as solid and as a 10 mM solution in DMSO.
  • Purity listed as 95.0%.
  • Molecular formula C7H10O3; molecular weight 142.15 g/mol.
  • Solubility ≥ 2.5 mg/mL (≈17.59 mM) in recommended solvents.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.