Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-benzenediol, 5-methyl-, hydrate | 6153-39-5 | MFCD00149092 | 95.0% | 142.15 g/mol | C7H10O3 | 25 G
SDP

Catalog No. 5000304136
Encompass_Preferred

5-Methylresorcinol monohydrate is a research reagent and tyrosinase inhibitor reported to exhibit cytotoxic and anxiolytic activity and to cross the blood-brain barrier. It is supplied as a monohydrate solid suitable for biochemical assays, screening, and preparative work, and is also available in a ready-to-use DMSO solution format for assays.

  • Formula C7H10O3; molecular weight 142.15 g/mol.
  • CAS number 6153-39-5.
  • Purity 95.0% (typical).
  • Physical form: off-white to light-yellow solid (monohydrate).
  • Available pack sizes include 5 g, 10 g, 25 g, and larger.
  • Also available as a 10 mM solution in DMSO (1 mL) for immediate use.
  • Storage: store solid at 4°C, protect from moisture; store solutions frozen for long-term stability.
  • Applications include tyrosinase inhibition assays, cytotoxicity studies, and neurological research.

Catalog No. 50-003-04136 Supplier Medchemexpress LLC Supplier No. 25GHYW015715
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methylresorcinol monohydrate is a research reagent and tyrosinase inhibitor reported to exhibit cytotoxic and anxiolytic activity and to cross the blood-brain barrier. It is supplied as a monohydrate solid suitable for biochemical assays, screening, and preparative work, and is also available in a ready-to-use DMSO solution format for assays.

  • Formula C7H10O3; molecular weight 142.15 g/mol.
  • CAS number 6153-39-5.
  • Purity 95.0% (typical).
  • Physical form: off-white to light-yellow solid (monohydrate).
  • Available pack sizes include 5 g, 10 g, 25 g, and larger.
  • Also available as a 10 mM solution in DMSO (1 mL) for immediate use.
  • Storage: store solid at 4°C, protect from moisture; store solutions frozen for long-term stability.
  • Applications include tyrosinase inhibition assays, cytotoxicity studies, and neurological research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.