Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzenedisulfonamide 98.0+%, TCI America™
SDP

Catalog No. B4917200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4917200MG 200 mg
B49171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4917200MG Supplier TCI America Supplier No. B4917200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3701-01-7
Molecular Formula C6H8N2O4S2
Molecular Weight (g/mol) 236.26
MDL Number MFCD00025383
InChI Key HUYYFHGIHVULSU-UHFFFAOYSA-N
PubChem CID 3090044
ChEBI CHEBI:35085
IUPAC Name benzene-1,3-disulfonamide
SMILES C1=CC(=CC(=C1)S(=O)(=O)N)S(=O)(=O)N

Specifications

Melting Point 235°C
Color White
Quantity 200 mg
Formula Weight 236.26
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Benzenedisulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.