Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzenedithiol, 97%

Catalog No. AAL1943906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1943906 5 g
AAL1943903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1943906 Supplier Thermo Scientific Chemicals Supplier No. L1943906
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 626-04-0 | C6H6S2 | 142.23 g/mol

1,3-Benzenedithiol is used to improve power conversion efficiency in cadmium selenide nanorods/poly(3-hexylthiophene) hybrid solar cells. It acts as an intermediate as well as involved in the preparation of 1,3-bis(-styrylthio)-benzol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzenedithiol is used to improve power conversion efficiency in cadmium selenide nanorods/poly(3-hexylthiophene) hybrid solar cells. It acts as an intermediate as well as involved in the preparation of 1,3-bis(-styrylthio)-benzol.

Solubility
Miscible with acetonitrile.

Notes
Incompatible with strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-04-0
Molecular Formula C6H6S2
Molecular Weight (g/mol) 142.23
MDL Number MFCD00004842
InChI Key ZWOASCVFHSYHOB-UHFFFAOYSA-N
Synonym 1,3-benzenedithiol, 1,3-dimercaptobenzene, dithioresorcin, dithioresorcinol, 1,3-benzedithiol, acmc-20ap5g, ksc357g0j
PubChem CID 522062
IUPAC Name benzene-1,3-dithiol
SMILES SC1=CC(S)=CC=C1

Specifications

Density 1.24
Boiling Point 244°C
Flash Point >112°C (234°F)
Odor Stench
Refractive Index 1.6625
Quantity 5 g
Beilstein 1634412
Sensitivity Air sensitive
Solubility Information Miscible with acetonitrile.
Formula Weight 142.24
Percent Purity 97%
Chemical Name or Material 1,3-Benzenedithiol
Show More Show Less
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 210-925-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.