Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-benzodioxol-5-ol, 7-methoxy- | 50901-34-3 | 96.0% | 168.15 g/mol | C8H8O4 | 1 ML
SDP

Catalog No. 5000210261
Encompass_Preferred

Antioxidant agent-4 (1,3-benzodioxol-5-ol, 7-methoxy-) is a small-molecule antioxidant reported to have antilipid peroxidative activity and is supplied for research use as solids and as a DMSO solution.

  • Antilipid peroxidative activity.
  • CAS number 50901-34-3.
  • Purity 96.0%.
  • Chemical formula C8H8O4 and molecular weight 168.15 g/mol.
  • High solubility in DMSO (100 mg/mL; ultrasonic may be required).
  • Storage: solid at 4°C under nitrogen; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-10261 Supplier Medchemexpress LLC Supplier No. HYW06842010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antioxidant agent-4 (1,3-benzodioxol-5-ol, 7-methoxy-) is a small-molecule antioxidant reported to have antilipid peroxidative activity and is supplied for research use as solids and as a DMSO solution.

  • Antilipid peroxidative activity.
  • CAS number 50901-34-3.
  • Purity 96.0%.
  • Chemical formula C8H8O4 and molecular weight 168.15 g/mol.
  • High solubility in DMSO (100 mg/mL; ultrasonic may be required).
  • Storage: solid at 4°C under nitrogen; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.