Learn More
Medchemexpress LLC 1,3-benzodioxole-5-carboxylic acid, 4-methyl- | 162506-58-3 | 96.3% | 180.16 | 5 G
SDP
Catalog No. 5000404535
Encompass_Preferred
4-Methyl-1,3-benzodioxole-5-carboxylic acid is a drug intermediate for the synthesis of various active compounds.
  • Used as a drug intermediate for synthesis of various active compounds.
  • Available in solid appearance, with a color ranging from white to off-white.
  • Provided with a data sheet, COA, SDS, and handling instructions for documentation.
  • Multiple quantities are available for purchase.
  • Shipping is at room temperature in the continental US; may vary elsewhere.
  • Storage recommendations: powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 6 months, -20°C for 1 month.
  • For research use only.
  • Free samples are offered.
Catalog No. 50-004-04535 Supplier Medchemexpress LLC Supplier No. HY414055G
4-Methyl-1,3-benzodioxole-5-carboxylic acid is a drug intermediate for the synthesis of various active compounds.
  • Used as a drug intermediate for synthesis of various active compounds.
  • Available in solid appearance, with a color ranging from white to off-white.
  • Provided with a data sheet, COA, SDS, and handling instructions for documentation.
  • Multiple quantities are available for purchase.
  • Shipping is at room temperature in the continental US; may vary elsewhere.
  • Storage recommendations: powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 6 months, -20°C for 1 month.
  • For research use only.
  • Free samples are offered.