Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzodioxole, 99%

Catalog No. AC170222500
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC170222500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC170222500 Supplier Thermo Scientific Chemicals Supplier No. 170222500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

CAS: 274-09-9 | C7H6O2 | 122.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 274-09-9
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00005818
InChI Key FTNJQNQLEGKTGD-UHFFFAOYSA-N
Synonym 1,2-methylenedioxybenzene, benzodioxole, benzo d 1,3 dioxole, 1,3-dioxaindan, 3,4-methylenedioxybenzene, 2h-1,3-benzodioxole, 1,2-methylenedioxy benzene, 1,3-dioxindan, methylenedioxybenzene, o-methylenedioxy benzene
PubChem CID 9229
ChEBI CHEBI:38732
IUPAC Name 1,3-benzodioxole
SMILES C1OC2=CC=CC=C2O1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -18.0°C
Color Brown to Yellow
Density 1.0600g/mL
Boiling Point 172.0°C to 173.0°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5380 to 1.5400
Quantity 250 g
Beilstein 19,2
Specific Gravity 1.06
Solubility Information Solubility in water: 0.2% (25 c). Other solubilities: miscible with most common organic solvents
Formula Weight 122.12
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 1, 3-Benzodioxole, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
EINECSNumber 205-992-0
RTECSNumber DA5600000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.