Learn More
Strem, An Ascensus Company CAS# 141556-45-8. 1g. 1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride, min. 97%. MFCD02684541
SDP
Catalog No. 5090108936
Encompass
CAS# 141556-45-8. 1g. 1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride, min. 97%. MFCD02684541. Molecular Weight: 340.90. Molecular Formula: C21H25N2+Clae. Color/form: off-white to yellow pwdr. Strem# 07-0299. http://www.strem.com/catalog/v/07-0299/
Catalog No. 50-901-08936 Supplier Strem, An Ascensus Company Supplier No. 0702991G
CAS# 141556-45-8. 1g. 1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride, min. 97%. MFCD02684541. Molecular Weight: 340.90. Molecular Formula: C21H25N2+Clae. Color/form: off-white to yellow pwdr. Strem# 07-0299. http://www.strem.com/catalog/v/07-0299/