Learn More
Strem, An Ascensus Company CAS# 141556-42-5. 500mg. 1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene, min. 98%. MFCD09265338
SDP
Catalog No. 5090109040
Encompass
CAS# 141556-42-5. 500mg. 1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene, min. 98%. MFCD09265338. Molecular Weight: 304.43. Molecular Formula: C21H24N2. Color/form: white to off-white pwdr. Strem# 07-0600. http://www.strem.com/catalog/v/07-0600/
Catalog No. 50-901-09040 Supplier Strem, An Ascensus Company Supplier No. 070600500MG
Add to Cart
Add to Cart
CAS# 141556-42-5. 500mg. 1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene, min. 98%. MFCD09265338. Molecular Weight: 304.43. Molecular Formula: C21H24N2. Color/form: white to off-white pwdr. Strem# 07-0600. http://www.strem.com/catalog/v/07-0600/