Learn More
Strem, An Ascensus Company 2g. 1,3-Bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene[2-(i-propoxy)-5-(N,N-dimethyl aminosulfonyl)phenyl]methyleneruthenium(II) dichloride (resin supported) Zhan Catalyst II. MFCD08459353
SDP
Catalog No. 5090111753
Encompass
2g. 1,3-Bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene[2-(i-propoxy)-5-(N,N-dimethyl aminosulfonyl)phenyl]methyleneruthenium(II) dichloride (resin supported) Zhan Catalyst II. MFCD08459353. Molecular Weight: >1000. Molecular Formula: . Color/form: black solid; Loading: 0.5 mmol/g Strem# 44-0083. http://www.strem.com/catalog/v/44-0083/
Catalog No. 50-901-11753 Supplier Strem, An Ascensus Company Supplier No. 4400832G
2g. 1,3-Bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene[2-(i-propoxy)-5-(N,N-dimethyl aminosulfonyl)phenyl]methyleneruthenium(II) dichloride (resin supported) Zhan Catalyst II. MFCD08459353. Molecular Weight: >1000. Molecular Formula: . Color/form: black solid; Loading: 0.5 mmol/g Strem# 44-0083. http://www.strem.com/catalog/v/44-0083/