Learn More
Strem, An Ascensus Company CAS# 244187-81-3. 1g. 1,3-Bis(2,6-di-i-propylphenyl)imidazol-2-ylidene, min. 98%. MFCD07781991
SDP
Catalog No. 501385337
Encompass
CAS# 244187-81-3. 1g. 1,3-Bis(2,6-di-i-propylphenyl)imidazol-2-ylidene, min. 98%. MFCD07781991. Molecular Weight: 388.59. Molecular Formula: C27H36N2. Color/form: white to off-white pwdr. Strem# 07-0595. http://www.strem.com/catalog/v/07-0595/
Catalog No. 50-138-5337 Supplier Strem, An Ascensus Company Supplier No. 0705951G
CAS# 244187-81-3. 1g. 1,3-Bis(2,6-di-i-propylphenyl)imidazol-2-ylidene, min. 98%. MFCD07781991. Molecular Weight: 388.59. Molecular Formula: C27H36N2. Color/form: white to off-white pwdr. Strem# 07-0595. http://www.strem.com/catalog/v/07-0595/