Learn More
Strem, An Ascensus Company CAS# 852445-83-1. 1g. 1,3-Bis(2,6-di-isopropylphenyl)imidazol-2-ylidenegold(I) chloride, 95%. MFCD09839143
SDP
Catalog No. 5090113384
Encompass
CAS# 852445-83-1. 1g. 1,3-Bis(2,6-di-isopropylphenyl)imidazol-2-ylidenegold(I) chloride, 95%. MFCD09839143. Molecular Weight: 621.01. Molecular Formula: C27H36AuClN2. Color/form: white pwdr. Strem# 79-0200. http://www.strem.com/catalog/v/79-0200/
Catalog No. 50-901-13384 Supplier Strem, An Ascensus Company Supplier No. 7902001G
CAS# 852445-83-1. 1g. 1,3-Bis(2,6-di-isopropylphenyl)imidazol-2-ylidenegold(I) chloride, 95%. MFCD09839143. Molecular Weight: 621.01. Molecular Formula: C27H36AuClN2. Color/form: white pwdr. Strem# 79-0200. http://www.strem.com/catalog/v/79-0200/