Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(2,6-diisopropylphenyl)imidazolidin-2-ylidene 98.0+%, TCI America™
SDP

Catalog No. B35061G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35061G Supplier TCI America Supplier No. B35061G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 258278-28-3
Molecular Formula C27H38N2
Molecular Weight (g/mol) 390.615
MDL Number MFCD10567379
InChI Key XZDYFCGKKKSOEY-UHFFFAOYSA-N
Synonym SIPr
PubChem CID 10883669
IUPAC Name 1,3-bis[2,6-di(propan-2-yl)phenyl]-4,5-dihydro-2H-imidazol-1-ium-2-ide
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N2CC[N+](=[C-]2)C3=C(C=CC=C3C(C)C)C(C)C

Specifications

Color White
Quantity 1 g
Formula Weight 390.62
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(2,6-diisopropylphenyl)imidazolidin-2-ylidene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.