Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(2-hydroxyethylsulfonyl)-2-propanol 93.0+%, TCI America™
SDP

Catalog No. B269925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B269925G 25 g
B2699500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B269925G Supplier TCI America Supplier No. B269925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67006-34-2
Molecular Formula C7H16O7S2
Molecular Weight (g/mol) 276.318
InChI Key CVFXDAXIZHJZAH-UHFFFAOYSA-N
PubChem CID 11437335
IUPAC Name 1,3-bis(2-hydroxyethylsulfonyl)propan-2-ol
SMILES C(CS(=O)(=O)CC(CS(=O)(=O)CCO)O)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 276.32
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(2-hydroxyethylsulfonyl)-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.