Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea 98.0+%, TCI America™
SDP

Catalog No. B34521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34521G 1 g
B3452200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34521G Supplier TCI America Supplier No. B34521G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1060-92-0
Molecular Formula C17H8F12N2S
Molecular Weight (g/mol) 500.306
InChI Key RWXWQJYJWJNJNW-UHFFFAOYSA-N
PubChem CID 2803772
IUPAC Name 1,3-bis[3,5-bis(trifluoromethyl)phenyl]thiourea
SMILES C1=C(C=C(C=C1C(F)(F)F)NC(=S)NC2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 1 g
Formula Weight 500.31
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.