Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Bis(3-Glycidyloxypropyl)Tetramethyldisiloxane

Catalog No. 14845100MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-845-100MLF 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 14-845-100MLF Supplier Sigma Aldrich Supplier No. 14845100MLF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3-Bis(3-glycidyloxypropyl)tetramethyldisiloxane can be used:As a starting material to synthesize functional poly(siloxane amine)s via an amine-epoxy click polymerization for material and biological applications.To prepare polymer electrolyte membranes (PEM) for fuel cell applications.

Specifications

CAS 40372-72-3
Density 1.08 g/mL at 20°C
Molecular Formula C18H42O6S4Si2
Refractive Index n20/D 1.452
Linear Formula C18H42O6S4Si2
MDL Number MFCD00053751
Quantity 50 mL
Molecular Weight (g/mol) 538.95 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.