Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(4-hydroxyphenoxy)benzene 98.0+%, TCI America™
SDP

Catalog No. B14845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14845G 5 g
B14841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14845G Supplier TCI America Supplier No. B14845G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 126716-90-3
Molecular Formula C18H14O4
Molecular Weight (g/mol) 294.306
MDL Number MFCD00142505
InChI Key CJLPIPXJJJUBIV-UHFFFAOYSA-N
Synonym Resorcinol Bis(4-hydroxyphenyl) Ether
PubChem CID 626311
IUPAC Name 4-[3-(4-hydroxyphenoxy)phenoxy]phenol
SMILES C1=CC(=CC(=C1)OC2=CC=C(C=C2)O)OC3=CC=C(C=C3)O

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 5 g
Formula Weight 294.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(4-hydroxyphenoxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.