Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-bis(4-hydroxyphenyl)-4-methyl-5-[4-(2-piperidinylethoxy)phenol]-1H-pyrazole dihydrochloride | 911295-24-4 | MFCD20524909 | 99.5% | 542.50 g/mol | C29H33Cl2N3O3 | 25 MG
SDP

Supplier:  Medchemexpress LLC HY10345425MG

Encompass_Preferred

MPP dihydrochloride is a selective estrogen receptor α (ERα) antagonist used as a research reagent to probe ERα-mediated signaling and apoptosis in cellular and biochemical assays. It is supplied as a high-purity powder for in vitro studies, with recommended sealed, low-temperature storage for solutions.

  • Selective ERα antagonist with >200-fold selectivity over ERβ.
  • High purity suitable for biochemical and cell-based assays.
  • Molecular formula C29H33Cl2N3O3; molecular weight 542.50 g/mol.
  • Supplied in small research quantities (25 mg) as a white powder.
  • Recommended storage: 4°C sealed; in solution, -80°C for long-term stability.

Catalog No. 50-002-45131


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.