Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-bis(carboxyethoxy)-2,2-bis(carboxyethoxy)propane | 35638-19-8 | >98% | C17H28O12 | 5 G
SDP

Catalog No. 5000353048
Encompass_Preferred

1,3-bis(carboxyethoxy)-2,2-bis(carboxyethoxy)propane is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Purity >98%
  • Solid appearance
  • White to off-white color
  • Molecular weight: 424.40

Catalog No. 50-003-53048 Supplier Medchemexpress LLC Supplier No. HY1405275G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-bis(carboxyethoxy)-2,2-bis(carboxyethoxy)propane is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Purity >98%
  • Solid appearance
  • White to off-white color
  • Molecular weight: 424.40

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.