Learn More
Medchemexpress LLC 1,3-bis(carboxyethoxy)-2,2-bis(carboxyethoxy)propane | 35638-19-8 | C17H28O12 | 10 G
SDP
Catalog No. 5000356902
Encompass_Preferred
1,3-bis(carboxyethoxy)-2,2-bis(carboxyethoxy)propane is a PEG-based PROTAC linker for synthesizing PROTACs. PROTACs use two ligands connected by a linker, one for an E3 ubiquitin ligase and another for a target protein, to exploit the intracellular ubiquitin-proteasome system for selective protein degradation. It is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-56902 Supplier Medchemexpress LLC Supplier No. HY14052710G
1,3-bis(carboxyethoxy)-2,2-bis(carboxyethoxy)propane is a PEG-based PROTAC linker for synthesizing PROTACs. PROTACs use two ligands connected by a linker, one for an E3 ubiquitin ligase and another for a target protein, to exploit the intracellular ubiquitin-proteasome system for selective protein degradation. It is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only