Learn More
Medchemexpress LLC 1,3-bis(carboxyethoxy)-2,2-bis(carboxyethoxy)propane | 35638-19-8 | C17H28O12 | 250 MG
SDP
Catalog No. 5000352777
Encompass_Preferred
1,3-bis(carboxyethoxy)-2,2-bis(carboxyethoxy)propane is a PEG-based PROTAC linker used in the synthesis of PROTACs. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • PROTACs contain two ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase, the other targets a specific protein
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52777 Supplier Medchemexpress LLC Supplier No. 250MGHY140527
1,3-bis(carboxyethoxy)-2,2-bis(carboxyethoxy)propane is a PEG-based PROTAC linker used in the synthesis of PROTACs. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • PROTACs contain two ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase, the other targets a specific protein
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins