Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Bis(Cbz-amino)-bicyclo[1.1.1]pentane

Catalog No. PHB00013 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PHB00013 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PHB00013 Supplier Sigma Aldrich Supplier No. PHB00013100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-hydroxyphenethyl alcohol reacts with triethyl or trimethyl aluminium to yield cyclic products and organoaluminim oligomers
  • Inelastic electron tunnelling spectra of 2-hydroxyphenethyl alcohol adsorbed on thin-film aluminium and magnesium oxide has been investigated

Specifications

CAS 1566649-44-2
MDL Number MFCD27987021
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.